missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgE Monoclonal Antibody (Ige21), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£160.00 - £340.00
Specifications
| Antigen | Human IgE |
|---|---|
| Clone | Ige21 |
| Concentration | 0.2 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055019
|
Invitrogen™
H077T03R04-A |
100 Tests |
£340.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055316
|
Invitrogen™
H077T02R04-A |
25 Tests |
£160.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgE Monoclonal antibody specifically detects IgE in Human samples. It is validated for Flow CytometrySpecifications
| Human IgE | |
| 0.2 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01854 | |
| 3497 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| Ige21 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; Immunoglobulin E; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; ImmunoglobulinE; Km | |
| IgG1 κ | |
| Affinity chromatography | |
| IGHE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title