missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgE Uncoated ELISA Kit

Product Code. 15530957
Change view
Click to view available options
Quantity:
20 x 96 Tests
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15530957 20 x 96 Tests Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15530957 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' 885061077

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ELISA

The Human Immunoglobulin E (Hu IgE) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgE. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.
TRUSTED_SUSTAINABILITY

Spezifikation

Assay Range 7.8 to 500 ng/mL
Assay Sensitivity 7.8 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3497
Kit Contents Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, Biotin-conjugated antibody, Standard: Human IgE protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Avidin-HRP, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Quantity 20 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 10 μL, Serum 10 μL
Synonym Immunoglobulin E
Target Human IgE
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.