missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE, Goat anti-Rat, HRP, Secondary Antlibody, Abnova™
Goat Secondary Antibody
Brand: Abnova PAB29751.500ug
This item is not returnable.
View return policy
Specifications
| ELISA, Immunohistochemistry, Western Blot | |
| ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.5-5 ug/mL) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Goat | |
| 500 μg | |
| Secondary | |
| Store at 4°C. | |
| Liquid |
| HRP | |
| In 50 mM HEPES-Saline with stabilizer (0.002 % Thimerosal) | |
| Rat IgE whole molecule. | |
| RUO | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction