missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Antibody (BE5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-470AF647
This item is not returnable.
View return policy
Description
IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| IgE | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGHE | |
| Purified human IgE. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3497 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| BE5 | |
| Flow Cytometry, ELISA | |
| IgE, immunoglobulin heavy constant epsilon | |
| Mouse | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| The antibody BE5 reacts with human IgE; it recognizes an epitope different from the ones recognized by 4G7 and 4H10 antibodies to IgE. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction