Learn More
IgE Antibody (3H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11623-0.025mg
 View more versions of this product
Product Code. 18601919
  • £149.00 / 25µg
Description
IgE Monoclonal antibody specifically detects IgE in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgE
Monoclonal
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Equine
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
3H10
Western Blot, ELISA 1:1000 - 1:10000, Immunohistochemistry, Immunohistochemistry-Frozen
IgE, immunoglobulin heavy constant epsilon
Equine IgE
0.025 mg
Adaptive Immunity, Immunology
3497
Store at -20°C. Avoid freeze-thaw cycles.
IgG1