missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgD Monoclonal Antibody (IA6-2), Brilliant Violet™ 650, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£143.00 - £331.00
Specifications
| Antigen | Human IgD |
|---|---|
| Clone | IA6-2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17847032
|
Invitrogen™
416-9868-41 |
25 Tests |
£143.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17886912
![]() |
Invitrogen™
416-9868-42 |
100 Tests |
£331.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
| Human IgD | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| IGHD; immunoglobulin heavy constant delta | |
| IGHD | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| IA6-2 | |
| Flow Cytometry | |
| Brilliant Violet 650 | |
| Mouse | |
| Human | |
| P01880 | |
| 3495 | |
| IgG2a κ | |
| Affinity chromatography | |
| IGHD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title