missing translation for 'onlineSavingsMsg'
Learn More

IgD Monoclonal Antibody (11-26c (11-26)), Super Bright™ 702, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

£133.00 - £331.00

Specifications

Antigen IgD
Clone 11-26c (11-26)
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15983774
View Documents
Invitrogen
67-5993-80
25 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15993774
GREENER_CHOICE
View Documents
Invitrogen™
67-5993-82
100 μg
£331.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells.

This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Specifications

Specifications

IgD
0.2 mg/mL
Monoclonal
Liquid
RUO
380797
IgG2a κ
Affinity chromatography
Antibody
11-26c (11-26)
Flow Cytometry
Super Bright 702
Rat
Mouse
Ighd
Primary
4° C, store in dark, DO NOT FREEZE!
Ighd
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.