Learn More
IgD Monoclonal Antibody (11-26c (11-26)), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 64-5993-82
Additional Details : Weight : 0.01000kg
Description
Description: The 8A7 monoclonal antibody reacts with the human BAFF receptor, also known as B lymphocyte stimulator (BLyS protein) receptor, which is expressed on B cells. The ligand for this receptor, BAFF (B cell-activating factor of the TNF family) is a B cell survival factor and regulates CD21/35 and CD23 expression. Interaction of this ligand with its receptor causes elevated CD21/35 and CD23 expression, whereas receptor blockade has been shown to down-modulate expression. Applications Reported: This 8A7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 8A7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
IgD | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P01881 | |
Ighd | |
Affinity chromatography | |
RUO | |
380797 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
11-26c (11-26) | |
Super Bright 645 | |
Ighd | |
IgD; Igh-5; IGHD; immunoglobulin heavy constant delta | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |