Learn More
Invitrogen™ Mouse IgD Monoclonal Antibody (10.4.22), Functional Grade, eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen™ 16-5924-85
Description
Description: This 10.4.22 monoclonal antibody reacts with the a allotype of mouse IgD, which is expressed by mature peripheral B cells. This antibody has been reported to activate B cells via engagement of the IgD receptor. The use of anti-IgM as a polyclonal B cell activator is problematic due to interference from high levels of circulating serum IgM. Crossblocking studies suggest that the 10.4.22 antibody recognizes a different epitope from 11-26c. Applications Reported: This 10.4.22 antibody has been reported for use in functional assays. Applications Tested: This 10.4.22 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Specifications
| Mouse IgD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P01881 | |
| Ighd | |
| Affinity chromatography | |
| RUO | |
| 380797 | |
| 4°C | |
| Liquid |
| Functional Assay | |
| 10.4.22 | |
| Functional Grade | |
| Ighd | |
| IgD; Igh-5; IGHD; immunoglobulin heavy constant delta | |
| Mouse | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.