missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGBP1 Antibody (OTI4F6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71009H
This item is not returnable.
View return policy
Description
IGBP1 Monoclonal antibody specifically detects IGBP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF
Specifications
| IGBP1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| OTI4F6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, CyTOF-ready | |
| ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16 | |
| Full length human recombinant protein of human IGBP1 (NP_001542) produced in HEK293T cell. | |
| 0.1 mL | |
| Protein Phosphatase, Signal Transduction | |
| 3476 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction