missing translation for 'onlineSavingsMsg'
Learn More

IGBP1 Antibody, Novus Biologicals™

Product Code. 18430081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430081 25 μL 25µL
18229046 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18430081 Supplier Novus Biologicals Supplier No. NBP18312525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

IGBP1 Polyclonal specifically detects IGBP1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGBP1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16
Gene Symbols IGBP1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PELFETGRQLLDEVEVATEPAGSRIVQEKVFKGLDLLEKAAEMLSQLDLFSRNEDLEEIASTDLKYLLVPAFQGALTMKQVNPSKRLDHLQRAREHFINYLTQCHCYHVAEFELP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3476
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.