missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgA Rat Uncoated ELISA Kit

For quantitative detection of rat IgA.

Brand:  Invitrogen™ 88-50480-77

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15530917

  • £524.00 / Pack of 20
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Includes: Capture Antibody Concentrate

Description

Description

The Rat Total Immunoglobulin G (IgG) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rat IgA Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

16ng/mL
Rat
20 x 96 Tests
Immunoglobulin A
Cell Culture Supernatant, Plasma, Serum
ELISA
2°C to 8°C
IgA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only