missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig, λ Light Chain Mouse anti-Human, BUV563, Clone: 1-155-2, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751017
This item is not returnable.
View return policy
Description
Ig, λ Light Chain Monoclonal antibody specifically detects Ig, λ Light Chain in Human samples. It is validated for Flow Cytometry
Specifications
| Ig, λ Light Chain | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| IGL; IGL@; Ig λ; Immunoglobulin lambda | |
| Human IgA1-λ myeloma Protein | |
| RUO | |
| Human | |
| IgG1 λ |
| Flow Cytometry | |
| 1-155-2 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction