missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig Light Chain Antibody (RM129), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77396
This item is not returnable.
View return policy
Description
Ig Light Chain Monoclonal antibody specifically detects Ig Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Ig Light Chain | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 μg/mL-2 μg/mL, ELISA 0.1 μg/mL - 0.5 μg/mL, Immunohistochemistry 0.5 μg/mL - 2 μg/mL, Immunocytochemistry/ Immunofluorescence 0.5 μg/mL - 2 μg/mL | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| RM129 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Human IgG | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction