missing translation for 'onlineSavingsMsg'
Learn More

Ig Light Chain Antibody (RM129), Biotin, Novus Biologicals™

Product Code. 18338174 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338174 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338174 Supplier Novus Biologicals Supplier No. NBP318532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Ig Light Chain Monoclonal antibody specifically detects Ig Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig Light Chain
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
Clone RM129
Concentration 1 mg/ml
Conjugate Biotin
Dilution Flow Cytometry 0.5 to 2 μg/mL, ELISA 0.1 to 0.5 μg/mL, Immunohistochemistry 0.5to 2 μg/mL, Immunocytochemistry/Immunofluorescence 0.5to 2 μg/mL, Sandwich ELISA
Formulation 50% Glycerol, PBS, 1% BSA
Host Species Rabbit
Immunogen Human IgG
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.