missing translation for 'onlineSavingsMsg'
Learn More

Ig Lambda C Domain/IGLC2 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30734100 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734100 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734100 Supplier Novus Biologicals Supplier No. AF6525D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Ig Lambda C Domain/IGLC2 Polyclonal antibody specifically detects Ig Lambda C Domain/IGLC2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig Lambda C Domain/IGLC2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias IGLC, immunoglobulin lambda constant 2 (Kern-Oz- marker), MGC20392, MGC45681
Host Species Goat
Immunogen E. coli-derived recombinant human IGLC2, Gln2-Ser106, Accession # P0CG05
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3538
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.