missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig kappa light chain Mouse anti-Human, Clone: Kap-56, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14289.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human Ig kappa light chain.
- High specificity
- High reactivity
Specifications
| Ig kappa light chain | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-2 ug/106 cells in 0.1 mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P01834 | |
| IGKC | |
| Human lymphocytes stimulated. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
| Kap-56 | |
| Mouse monoclonal antibody raised against native human Ig kappa light chain. | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3514 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction