missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig gamma-4 Chain C Region, Rabbit anti-Human, Clone: IGHG4/2042R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-79716-100UG
This item is not returnable.
View return policy
Description
Ig gamma-4 Chain C Region Monoclonal specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Ig gamma-4 Chain C Region | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P01861 | |
| IGHG4 | |
| Recombinant full-length human Ig gamma-4 Chain C Region protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| IGHG4/2042R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3503 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction