missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ig gamma-4 Chain C Region Antibody (IGHG4/1345), DyLight 680, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Ig gamma-4 Chain C Region |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | IGHG4/1345 |
| Conjugate | DyLight 680 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker) |
| Host Species | Mouse |
| Immunogen | Recombinant human Ig gamma-4 Chain C Region fragment (Uniprot: P01861) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?