missing translation for 'onlineSavingsMsg'
Learn More

Ig gamma-4 Chain C Region Antibody (IGHG4/1345), DyLight 550, Novus Biologicals™

Product Code. 18430755 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430755 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430755 Supplier Novus Biologicals Supplier No. NBP254461R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ig gamma-4 Chain C Region Monoclonal antibody specifically detects Ig gamma-4 Chain C Region in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig gamma-4 Chain C Region
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IGHG4/1345
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias IGHG4, mmunoglobulin heavy constant gamma 4, mmunoglobulin heavy constant gamma 4 (G4m marker)
Host Species Mouse
Immunogen Recombinant human Ig gamma-4 Chain C Region fragment (Uniprot: P01861)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3503
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.