missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFT88 Recombinant Rabbit Monoclonal Antibody (23GB4015), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | IFT88 |
|---|---|
| Clone | 23GB4015 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089318
![]() |
Invitrogen™
MA553670 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088664
|
Invitrogen™
MA553671 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFT88 Recombinant Monoclonal antibody specifically detects IFT88 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| IFT88 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q13099, Q61371 | |
| 21821, 305918, 8100 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4015 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AW552028; D13S1056E; DAF19; fb37b11; flexo; fxo; HGNC:20606; hTg737; Ift88; intraflagellar transport 88; intraflagellar transport 88 homolog; intraflagellar transport protein; intraflagellar transport protein 88; intraflagellar transport protein 88 homolog; MGC26259; orpk; osm-5; oval; ovl; polaris; polaris homolog; polaris; axonemal transport protein; probe hTg737 (polycystic kidney disease, autosomal recessive); recessive polycystic kidney disease protein Tg737; Recessive polycystic kidney disease protein Tg737 homolog; RP11-172H24.2; testicular tissue protein Li 93; tetratricopeptide repeat domain 10; tetratricopeptide repeat protein 10; TG737; Tg737 protein; Tg737Rpw; TgN(Imorpk)737Rpw; TgN737Rpw; TPR repeat protein 10; transgene insert site 737, insertional mutation, polycystic kidney disease; Ttc10; wu:fb37b11; xx:zah8127 | |
| A synthesized peptide derived from IFT88 (1-28AA). | |
| Primary | |
| -20°C | |
| IFT88 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title