missing translation for 'onlineSavingsMsg'
Learn More

IFT88 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18382240 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382240 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382240 Supplier Novus Biologicals Supplier No. NBP2866750.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFT88 Polyclonal specifically detects IFT88 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT88
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of IFT88
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8100
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.