missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Antibody (JE64-77), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32459
This item is not returnable.
View return policy
Description
IFT88 Monoclonal antibody specifically detects IFT88 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| IFT88 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| D13S1056ETPR repeat protein 10, DAF19, hTg737, intraflagellar transport 88 homolog (Chlamydomonas), intraflagellar transport protein 88 homolog, MGC26259, polaris homolog, probe hTg737 (polycystic kidney disease, autosomal recessive), Recessive polycystic kidney disease protein Tg737 homolog, tetratricopeptide repeat domain 10, Tetratricopeptide repeat protein 10, TG737Tg737, TTC10 | |
| Synthetic peptide within Human IFT88 aa 1-50/ 824. (Uniprot: Q13099) | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication | |
| 8100 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE64-77 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction