missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT81 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97511PCP
This item is not returnable.
View return policy
Description
IFT81 Polyclonal antibody specifically detects IFT81 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| IFT81 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| carnitine deficiency-associated gene expressed in ventricle 1, Carnitine deficiency-associated protein expressed in ventricle 1, CDV-1, CDV1carnitine deficiency-associated, expressed in ventricle 1, CDV1R, CDV-1R, intraflagellar transport 81 homolog (Chlamydomonas), intraflagellar transport protein 81 homolog, MGC102777, MGC4027 | |
| Produced in rabbits immunized with E. coli-derived Human IFT81 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 28981 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction