missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT74 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27334-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.Specifications
| IFT74 | |
| Polyclonal | |
| Unconjugated | |
| IFT74 | |
| CCDC2, CMG 1, CMG1, IFT74 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 67694, 80173 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BKE9, Q96LB3 | |
| Ift74 | |
| IFT74 Fusion Protein Ag26385 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction