missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT74 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35263-20ul
This item is not returnable.
View return policy
Description
IFT74 Polyclonal antibody specifically detects IFT74 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| IFT74 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:3000, ELISA | |
| Capillary morphogenesis gene 1 protein, CCDC2capillary morphogenesis protein 1, CMG1Coiled-coil domain-containing protein 2, CMG-1MGC111562, coiled-coil domain containing 2, FLJ22621, intraflagellar transport 74 homolog (Chlamydomonas), intraflagellar transport protein 74 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-372 of human IFT74 (NP_001092694.1).,, Sequence:, MASNHKSSAARPVSRGGVGLTGRPPSGIRPLSGNIRVATAMPPGTARPGSRGCPIGTGGVLSSQIKVAHRPVTQQGLTGMKTGTKGPQRQILDKSYYLGLLRSKISELTTEVNKLQKGIEMYNQENSVYLSYEKRAETLAVEIKELQGQLADYNMLVDKLNTNTEMEEVMNDYNMLKAQNDRETQSLDVIFTERQAKEKQIRSVEEEIEQEKQATDDIIKNMSFENQVKYLEMKTTNEKLLQELDTLQQQLDSQNMKKESLEAEIAHSQVKQEAVLLHEKLYELESHRDQMIAEDKSIGSPMEEREKLLKQIKDDNQEIASMERQLTDTKEKINQFIEEIRQLDMDLEEHQDPTNYGWKILEKNTGSFKKQV | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 80173 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction