Learn More
IFT52 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566920
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: EPLQLIQPQFE TPLPTLQPAV FPPSFRELPP PPLELFDLDE TFSSEKARLA QITNKCTEED LEFYVRKCGD ILGVTSKLPK DQQDA Highest antigen sequence identity to the following orthologs - mouse 95%, rat 27%.
Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia.Specifications
IFT52 | |
Polyclonal | |
Unconjugated | |
IFT52 | |
BC037708; C20orf9; CGI-53; Ift52; intraflagellar transport 52; intraflagellar transport 52 homolog; intraflagellar transport protein 52 homolog; NGD2; NGD5; Protein NGD5; Protein NGD5 homolog; RGD1311004 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51098 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y366 | |
IFT52 | |
Recombinant Human IFT52 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only