missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFT140 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559910
This item is not returnable.
View return policy
Description
Immunogen sequence: LWRLDQRGRV QGTPLLKHEY GKHLTHCIFR LPPPGEDLVQ LAKAAVSGDE KALDMFNWKK SSSGSLLKMG SHEGLLFFVS LMDGTVHYVD EKGKTTQVVS ADSTIQMLFY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.
Specifications
| IFT140 | |
| Polyclonal | |
| Unconjugated | |
| IFT140 | |
| AI661311; c305C8.4; c380F5.1; gs114; Ift140; intraflagellar transport 140; intraflagellar transport 140 homolog; intraflagellar transport protein 140 homolog; KIAA0590; LOW QUALITY PROTEIN: intraflagellar transport protein 140 homolog; mKIAA0590; MZSDS; SRTD9; Tce5; T-complex expressed gene 5; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2; WDTC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9742 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RY7 | |
| IFT140 | |
| Recombinant protein corresponding to Human IFT140. Recombinant protein control fragment (Product #RP-98798). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction