missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT122 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544136
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QADPAQKDTM LGKFYHFQRL AELYHGYHAI HRHTEDPFSV HRPETLFNIS Sequence homology: Human: 100%
IFT122 Polyclonal antibody specifically detects IFT122 in Human samples. It is validated for Western Blot.Specifications
IFT122 | |
Polyclonal | |
Unconjugated | |
IFT122 | |
C86139; CED; CED1; hypothetical protein zgc:77217; Ift122; intraflagellar transport 122; intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas); intraflagellar transport protein 122 homolog; sopb; SPG; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140; WDR10; WDR10p; WDR140; zgc:77217 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55764 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HBG6 | |
IFT122 | |
Synthetic peptide directed towards the C-terminal of human IFT122 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |