missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT122 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97508MFV500
This item is not returnable.
View return policy
Description
IFT122 Polyclonal antibody specifically detects IFT122 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IFT122 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| intraflagellar transport 122 homolog (Chlamydomonas), intraflagellar transport protein 122 homolog, SPGWD repeat-containing protein 140, WD repeat domain 10, WD repeat-containing protein 10, WDR10WDR10p, WDR140CED | |
| Produced in rabbits immunized with E. coli-derived Human IFT122 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55764 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction