missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ iFSP1
GREENER_CHOICE

Product Code. 18717936
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18717936

Brand: Tocris Bioscience™ 7162/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ferroptosis suppressor protein 1 (FSP1) inhibitor; ferroptosis inducer

iFSP1 is a ferroptosis inducer. Inhibits ferroptosis suppressor protein 1 (FSP1) and induces ferroptosis in GPX4 knockout cells. Sensitizes tumor cells to RSL3 (Cat. No. 6118) -induced ferroptosis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile
CAS 150651-39-1
Quantity 10 mg
Target Other Reductase Inhibitors
Molecular Formula C20H13N5
Formula Weight 1414
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.