missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFRG15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21825-1-AP-20UL
This item is not returnable.
View return policy
Description
One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.Specifications
| IFRG15 | |
| Polyclonal | |
| Unconjugated | |
| TOR1AIP2 | |
| IFRG15, interferon responsive gene 15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 163590 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H496 | |
| TOR1AIP2 | |
| IFRG15 Fusion Protein Ag16156 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction