Learn More
IFRD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566999
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: CLACLESVFSR FYGLGGSSTS PVVPASLHGL LSAALQAWAL LLTICPSTQI SHILDRQLPR LPQLLSSESV NLRIAAGETI ALLFE Highest antigen sequence identity to the following orthologs - mouse 84%, rat 85%.
IFRD2 is a protein coding gene.Specifications
IFRD2 | |
Polyclonal | |
Unconjugated | |
IFRD2 | |
1810034A24Rik; AA470234; AI838527; IFNRP; IFRD2; interferon related developmental regulator 2; interferon-related developmental regulator 2; interferon-related protein; Protein SKMC15; SKMc15; SM15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12894 | |
IFRD2 | |
Recombinant Human IFRD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only