missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12939-1-AP-20UL
This item is not returnable.
View return policy
Description
Tis7, also known as IFRD1, is a 442 amino acid containing membrane associated, non-nuclear intracellular protein belonging to the IFRD family. A regulatory protein expressed in a variety of tissues, Tis7 is known to play an important role in the regulation of gene activity in the proliferative and/or differentiative pathways induced by NGF. Reports suggest that Tis7 may be an autocrine factor that attenuates or amplifies the initial ligand induced signal and might also augment the adaptive response by stimulating the production of differentiated enterocytes. Tis7 acts as a negative regulator of transcriptional activity and represses expression of genes involved in myogenesis, muscle maintece, and regeneration in a histone deacetylase dependent manner.Specifications
| IFRD1 | |
| Polyclonal | |
| Unconjugated | |
| IFRD1 | |
| IFRD1, PC4, TIS7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15982, 29596, 3475 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00458, P19182, P20695 | |
| IFRD1 | |
| IFRD1 Fusion Protein Ag3988 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction