missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IFRD1 Polyclonal antibody specifically detects IFRD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Regulatory Status | RUO |
| Research Discipline | Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?