missing translation for 'onlineSavingsMsg'
Learn More

Ifosfamide Impurity A, TRC

Product Code. 30388884
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388884 5 mg 5mg
30391889 25 mg 25mg
30405667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388884 Supplier Toronto Research Chemicals Supplier No. TRCI2650055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ifosfamide Impurity A
Analyte or Component Names Ifosfamide Impurity A
Shipping Condition Room Temperature
Molecular Formula C8 H17 Cl2 N2 O2 P
Synonym 3-(2-chloroethyl)-2-((3-chloropropyl)amino)-1,3,2-oxazaphosphinane 2-oxide
InChI Formula InChI=1S/C8H17Cl2N2O2P/c9-3-1-5-11-15(13)12(7-4-10)6-2-8-14-15/h1-8H2,(H,11,13)
Solution Type Neat
SMILES ClCCCNP1(=O)OCCCN1CCCl
IUPAC Name 3-(2-chloroethyl)-2-((3-chloropropyl)amino)-1,3,2-oxazaphosphinane 2-oxide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 275.112
Quantity 5 mg
Formula Weight 274.0405
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.