missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFNK Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5148977

Product Code. 30534549

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFNK
Polyclonal
Unconjugated
IFNK
Ifnk; IFN-kappa; IFNT1; INFE1; interferon; Interferon kappa; interferon, kappa; interferon-like protein; RP11-27J8.1; type I interferon kappa; UNQ6124/PRO20084
Sheep
Antigen affinity chromatography
RUO
387510
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q7TSL0
IFNK
E. coli-derived recombinant mouse IFN-kappa , Leu22-Lys199 (Cys174Tyr).
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.