missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNGR1 Mouse anti-Human, Clone: 1H3A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66704-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.Specifications
| IFNGR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15260 | |
| IFNGR1 | |
| IFNGR1 Fusion Protein Ag27085 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1H3A4 | |
| Unconjugated | |
| IFNGR1 | |
| CD119, CDw119, FLJ45734, IFN gamma R1, IFN gamma receptor 1, IFNGR, IFNGR1, interferon gamma receptor 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3459 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction