Learn More
Invitrogen™ IFNAR2 Monoclonal Antibody (MMHAR-2)

Description
This antibody neutralizes human IFN-alpha receptor, interacting with the extracellular domain. It binds to human IFN-alpha receptor with high affinity and blocks biological action of Type I IFNs. To measure bioactivity, interferon was titrated with the use of a cytopathic effect inhibition assay, where one neutralization unit is the amount of antiserum required to neutralize one unit of human interferon alpha to a 50% endpoint. In this antiviral assay, approximately one unit/mL of interferon was necessary to produce a cytopathic effect of 50%. Purity is >95% by SDS-PAGE and endotoxin level <1 EU/ug. Store at -20°C for up to 6 months. For long-term storage, store at -70°C for retention of full activity. Thaw product by incubation in cold water and aliquot into separate vials, adding 0.1% BSA for additional stability. Refreeze product using a dry ice or a dry ice/alcohol bath. Packaged by and distributed for PBL Assay Science.
Specifications
Specifications
| Antigen | IFNAR2 |
| Applications | Functional Assay, Neutralization |
| Classification | Monoclonal |
| Clone | MMHAR-2 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and no preservative |
| Gene | IFNAR2 |
| Gene Accession No. | P48551 |
| Gene Alias | AI747302; human interferon alpha/beta receptor; IFN alpha/betaRbeta; IFN R2; IFNABR; IFNalpha/beta R2; IFN-alpha/beta receptor 2; IFNalpha/betaR2; IFN-alpha/betaR2; IFN-alpha/betaRbeta; IFNalpha/beta-Rbeta; IFN-alpha-REC; IFNAR2; Ifnar-2; IFNARB; IFN-R; IFNR2; IFN-R-2; IMD45; interferon (alpha and beta) receptor 2; interferon (alpha, beta and omega) receptor 2; interferon alpha and beta receptor subunit 2; interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon receptor 2c; interferon-alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; type I interferon alpha/beta receptor 2a; type I interferon receptor 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.