Learn More
IFNAR1 Monoclonal Antibody (MAR1-5A3), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5945-050
Description
Description: The MAR1-5A3 monoclonal antibody reacts with mouse IFN alpha/beta receptor subunit 1 (IFNAR-1). The type I IFN receptor subunit 1 is co-expressed with IFNAR-2 on nearly all cells and make up the heterodimeric receptor complex that binds to all type I IFNs, IFN alpha and IFN beta. Type I IFNs are a group of structurally and functionally related proteins that demonstrate antiviral, antiparasitic, and antiproliferative activities. Applications Reported: The MAR1-5A3 antibody has been reported for use in neutralization of IFNAR1 bioactivity. Applications Tested: The Functional Grade Purified MAR1-5A3 antibody has been tested by LAL assay to verify low endotoxin levels, by bioassay for neutralization of mouse IFNAR-1bioactivity. The MAR1-5A3 antibody at 0.04 μg/mL has been found to inhibit by 50% the biological effects of 0.5 ng/mL mouse IFN-a2 (ND50), in an EMCV assay of L929 cell protection. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in Best Protocols® section. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
IFNAR1 Monoclonal antibody specifically detects IFNAR1 in Mouse samples. It is validated for Functional Assay, Neutralization
Specifications
| IFNAR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P33896 | |
| Mouse | |
| 50 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
| Functional Assay, Neutralization | |
| MAR1-5A3 | |
| Functional Grade | |
| Ifnar1 | |
| alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; IFNAR1; IFNBR; IFNR1; IFN-R-1; IFRC; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1 | |
| Affinity chromatography | |
| RUO | |
| 15975 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.