missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFNA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA570101
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Guinea Pig: 75%; Human: 100%; Mouse: 75%
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| IFNA7 | |
| Polyclonal | |
| Unconjugated | |
| IFNA7 | |
| Ifa7; IFNA7; IFNA-J; IFN-alpha 7; IFN-alpha-7; IFN-alphaJ; IFN-alpha-J1; interferon alpha 7; interferon alpha family, gene 7; interferon alpha-7; Interferon alpha-J; interferon alpha-J1; interferon, alpha 7; leIF J | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3444 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P01567 | |
| IFNA7 | |
| synthetic peptide directed towards the N terminal region of human IFNA7 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction