missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581605
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat IFNA5/IFNaG.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| IFNA5 | |
| Polyclonal | |
| PBS with no preservative | |
| 0 | |
| IFNA5 | |
| Recombinant Rat IFNA5/IFNaG protein. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| IFNA5 | |
| Ifa5; Ifna5; IFN-alpha-5; IFN-alphaG; INA5; INFA5; Interferon; interferon alpha 5; interferon alpha family, gene 5; interferon alpha-5; Interferon alpha-61; interferon alpha-G; interferon, alpha 5; leIF G | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 690894 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction