Learn More
Invitrogen™ Rat IFN-beta Recombinant Protein
Description
After receipt, this product should be kept at -20°C while lyophilized and at -70°C after reconstitution in sterile distilled water for retention of full activity. After reconstitution, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. After thawing, this product is stable for at least four weeks at 4°C. Further dilute only in buffers containing protein such as 0.1% bovine serum albumin (BSA).
Specifications
Specifications
| Accession Number | P01575, P70499 |
| For Use With (Application) | ELISA, Immunoprecipitation, Western Blot |
| Formulation | PBS with 0.125 M trehalose and no preservative |
| Gene ID (Entrez) | 15977, 24481 |
| Molecular Weight (g/mol) | 19.68 kDa |
| Name | Rat IFN-beta |
| Quantity | 1 x 10^5 units |
| Source | Hamster |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.