missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN omega Monoclonal Antibody (K16289_11E9), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | IFN omega |
|---|---|
| Clone | K16289_11E9 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30197363
|
Invitrogen™
MA555829 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30213106
|
Invitrogen™
MA556209 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GLLSRNTLVL LHQMRRISPF LCLKDRRDFR FPQEMVKGSQ LQKAHVMSVL HEMLQQIFSL FHTERSSAAW NMTLLDQLHT GLHQQLQHLE TCLLQVVGEG ESAGAISSPA LTLRRYFQGI RVYLKEKKYS DCAWEVVRME IMKSLFLSTN MQERLRSKDR
IFN omega Monoclonal antibody specifically detects IFN omega in Human samples. It is validated for Western BlotSpecifications
| IFN omega | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| IFN o; IFN ω; IFNo; IFNomega; IFN-omega 1, interferon omega-1; IFNW; IFNW1; IFNω; Interferon; Interferon alpha-II-1; Interferon o; Interferon OMEGA; interferon omega 1; interferon omega-1; Interferon ω; interferon, omega 1 | |
| Recombinant human IFNW1 (immunogen range is 31-190). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IFNW1 |
| K16289_11E9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05000 | |
| 3467 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title