missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-omega/IFNW1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | IFN-omega/IFNW1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18322554
|
Novus Biologicals
NBP3-03585-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18303074
|
Novus Biologicals
NBP3-03585-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-omega/IFNW1 Polyclonal specifically detects IFN-omega/IFNW1 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IFN-omega/IFNW1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-195 of human IFN-omega/IFNW1 (NP_002168.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 3467 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title