missing translation for 'onlineSavingsMsg'
Learn More

IFN-omega/IFNW1 Antibody (04), PE, Novus Biologicals™

Product Code. 18605067 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18605067 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605067 Supplier Novus Biologicals Supplier No. NBP306154PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-omega/IFNW1 Monoclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-omega/IFNW1
Applications ELISA
Classification Monoclonal
Clone 04
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IFN-omega/IFNW1 (Uniprot#: P05000; Met1-Ser195).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3467
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.