Learn More
IFN-γ Mouse, anti-Human, R718, Clone: B27, BD Horizon™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 566960
 View more versions of this product
Product Code. 17297744
  • ONLINE EXCLUSIVE PRICE
    £107.00
    £102.63 / 25 tests
    Save £4.37
    4% Off
Description
The B27 monoclonal antibody specifically binds to human interferon-γ (IFN-γ), a 14-18 kDa glycoprotein containing 143 amino acid residues. IFN-γ is a potent multifunctional cytokine produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ+ T cells. IFN-γ exerts its biological effects through specific binding to the high-affinity IFN-γ receptor complex comprised of IFN-γRα (CD119) and IFN-γRβ subunits. In addition to its antiviral effects, IFN-γ upregulates a number of lymphoid cell functions including the antimicrobial and anti-tumor responses of macrophages, NK cells, and neutrophils. In addition, IFN-γ influences the regulation of proliferation, differentiation, and effector responses of B cell and T cell subsets. These influences can involve IFN-γ's capacity to boost MHC class I and II expression by antigen-presenting cells as well as direct effects on B cells and T cells themselves. B27 is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-γ
Monoclonal
R718
25 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
B27