Learn More
IFN-γ Mouse anti-Human, PE, Clone: B27, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559327
Description
The B27 antibody reacts with human interferon-γ (IFN-γ. The immunogen used to generate the B27 hybridoma was E.coli-expressed recombinant human IFN-γ. This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development or are reported in the literature.
Intracellular Staining
Specifications
| IFN-γ | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| B27 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human E.coli-expressed IFN-γ Recombinant Protein | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.