missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCRD6 Monoclonal Antibody (196124)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA560975
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.
Specifications
| CCRD6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O00590 | |
| ACKR2 | |
| RBL-1 rat basophilic leukemia cell line transfected with human CC Chemokine Receptor D6, Met1-Ala384. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 196124 | |
| Unconjugated | |
| ACKR2 | |
| Ackr2; AI464239; Atypical chemokine receptor 2; C-C chemokine receptor D6; Ccbp2; CC-chemokine-binding receptor JAB61; CCR10; CCR10-related receptor; CCR9; chemokine (C-C motif) receptor 9; chemokine (C-C) receptor 9; chemokine binding protein 2; chemokine decoy receptor D6; chemokine receptor CCR-10; Chemokine receptor CCR-9; chemokine receptor D6; chemokine-binding protein 2; Chemokine-binding protein D6; CMKBR9; D6; D6 beta-chemokine receptor; hD6 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 1238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction