missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma, Rabbit anti-Human,Baboon, Clone: D9D10, Novus Biologicals™
Rabbit Monoclonal Antibody
£217.00 - £463.00
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | D9D10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, ELISA, Block/Neutralize |
| Applications | Western Blot, Flow Cytometry, ELISA, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18334411
|
Novus Biologicals
NBP2-81044-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18308330
|
Novus Biologicals
NBP2-81044-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human, Baboon samples. It is validated for Western Blot, Flow Cytometry, ELISA, Block/Neutralize.Specifications
| IFN-gamma | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| PBS | |
| 3458 | |
| FPLC-purified natural human IFN-gamma. Refer to the original article [PMID: 3114127] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| D9D10 | |
| Western Blot, Flow Cytometry, ELISA, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Baboon | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNG | |
| IgG κ | |
| Protein A purified | |
| Binds human IFN-gamma, a dimerized soluble cytokine that is the only member of the type II class of interferons. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title